CAS RN 1078166-57-0
CAS RN 1078166-57-0
IUPAC Name:
1-(4-fluorobenzoyl)-3-{[(6-methoxynaphthalen-2-yl)oxy]methyl}azetidine-3-carboxylic acid
Synonyms:
1-(4-fluorobenzoyl)-3-{[(6-methoxynaphthalen-2-yl)oxy]methyl}azetidine-3-carboxylic acid
Molecular Weight (g/mol):
409.41
Molecular Formula:
C23H20FNO5
InChi Key:
LWJGMYMNSNVCEM-UHFFFAOYSA-N
SMILES:
COC1=CC=C2C=C(OCC3(CN(C3)C(=O)C3=CC=C(F)C=C3)C(O)=O)C=CC2=C1
1
–
2
of
2
results
1
Tocris Bioscience™ PF 04418948

CAS: 1078166-57-0 Molecular Formula: C23H20FNO5 Molecular Weight (g/mol): 409.413
2
Medchem Express PF-04418948, MedChemExpress
MedChemExpress PF-04418948 is an orally active, potent and selective prostaglandin EP2 receptor antagonist with an IC50 of 16 nM.
