CAS RN 1079043-81-4
CAS RN 1079043-81-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Medchem Express Glycodeoxycholic acid monohydrate, MedChemExpress
MedChemExpress Glycodeoxycholic acid monohydrate is a nuclear receptor ligand.
