accessibility menu, dialog, popup

UserName

CAS RN 108-08-7

CH3H3CCH3H3C

CAS RN 108-08-7

IUPAC Name: 2,4-dimethylpentane
Synonyms: 2,4-dimethylpentane
Molecular Weight (g/mol): 100.21
Molecular Formula: C7H16
InChi Key: BZHMBWZPUJHVEE-UHFFFAOYSA-N
SMILES: CC(C)CC(C)C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results