accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108-52-1

CH3NH2NN

CAS RN 108-52-1

IUPAC Name: 4-methylpyrimidin-2-amine
Synonyms: 2-amino-4-methylpyrimidine
Molecular Weight (g/mol): 109.13
Molecular Formula: C5H7N3
InChi Key: GHCFWKFREBNSPC-UHFFFAOYSA-N
SMILES: CC1=CC=NC(N)=N1
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products