CAS RN 1080622-86-1
CAS RN 1080622-86-1
IUPAC Namn:
—
Synonymer:
—
Molekylvikt (g/mol):
—
Molekylär sammansättning:
—
InChi Key:
—
SMILES:
—
1
–
1
av
1
Resultat
1
CP-466722, MedChemExpress
MedChemExpress CP-466722 is a rapidly reversible inhibitor of ATM, with an IC50 of 4.1 μM, and has no effects on PI3K or closely related PI3K-like protein kinase (PIKK) family members.