accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108128-39-8

CH3NNOOHO

CAS RN 108128-39-8

IUPAC Name: 5-(furan-2-yl)-1-methyl-1H-pyrazole-3-carboxylic acid
Synonyms: 5-(furan-2-yl)-1-methylpyrazole-3-carboxylic acid
Molecular Weight (g/mol): 192.17
Molecular Formula: C9H8N2O3
InChi Key: XXEWHNSDJMBLBK-UHFFFAOYSA-N
SMILES: CN1N=C(C=C1C1=CC=CO1)C(O)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products