accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 108166-03-6

H3CNO

CAS RN 108166-03-6

IUPAC Name: 2-methylquinoline-6-carbaldehyde
Synonyms: 2-methylquinoline-6-carbaldehyde
Molecular Weight (g/mol): 171.2
Molecular Formula: C11H9NO
InChi Key: WAPQHBBEWPVBTO-UHFFFAOYSA-N
SMILES: CC1=CC=C2C=C(C=O)C=CC2=N1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products