accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109-56-8

H3CCH3NHOH

CAS RN 109-56-8

IUPAC Name: 2-[(propan-2-yl)amino]ethan-1-ol
Synonyms: 2-(isopropylamino)ethanol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: RILLZYSZSDGYGV-UHFFFAOYSA-N
SMILES: CC(C)NCCO

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products