accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109074-67-1

NHFFF

CAS RN 109074-67-1

IUPAC Name: 2-(trifluoromethyl)pyrrolidine
Synonyms: 2-(trifluoromethyl)pyrrolidine
Molecular Weight (g/mol): 139.12
Molecular Formula: C5H8F3N
InChi Key: VINAMCOZNJHNIH-UHFFFAOYNA-N
SMILES: FC(F)(F)C1CCCN1
Boiling Point 
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products