CAS RN 1092351-10-4
CAS RN 1092351-10-4
IUPAC Name:
8-(4-{[4-(4-chlorophenyl)piperazin-1-yl]sulfonyl}phenyl)-1-propyl-2,6-dihydro-1H-purine-2,6-dione
Synonyms:
8-{4-[4-(4-chlorophenyl)piperazin-1-ylsulfonyl]phenyl}-1-propylpurine-2,6-dione
Molecular Weight (g/mol):
527
Molecular Formula:
C24H23ClN6O4S
InChi Key:
OHVMMGQPJADDAJ-UHFFFAOYSA-N
SMILES:
CCCN1C(=O)N=C2N=C(N=C2C1=O)C1=CC=C(C=C1)S(=O)(=O)N1CCN(CC1)C1=CC=C(Cl)C=C1
1
–
1
of
1
results
1
Tocris Bioscience™ PSB 603

CAS: 1092351-10-4 Molecular Formula: C24H23ClN6O4S Molecular Weight (g/mol): 526.996