CAS RN 1092461-13-6
CAS RN 1092461-13-6
IUPAC Name:
1-[2,6-dichloro-3-(trifluoromethyl)phenyl]methanamine
Synonyms:
1-[2,6-dichloro-3-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol):
244.04
Molecular Formula:
C8H6Cl2F3N
InChi Key:
NDXOPDUEXPJASP-UHFFFAOYSA-N
SMILES:
NCC1=C(Cl)C=CC(=C1Cl)C(F)(F)F
1
–
1
of
1
results
1
2,6-Dichloro-3-(trifluoromethyl)benzylamine, 97%, Thermo Scientific™
CAS: 1092461-13-6 Molecular Formula: C8H6Cl2F3N Molecular Weight (g/mol): 244.04 MDL Number: MFCD11519352 InChI Key: NDXOPDUEXPJASP-UHFFFAOYSA-N Synonym: 2,6-dichloro-3-trifluoromethyl benzylamine,1-2,6-dichloro-3-trifluoromethyl phenyl methanamine,2,6-dichloro-3-trifluoromethyl benzylamine, jrd,2,6-dichloro-3-trifluoromethyl phenyl methanamine,2,6-dichloro-3-trifluoromethyl phenyl methylamine PubChem CID: 57365252 IUPAC Name: [2,6-dichloro-3-(trifluoromethyl)phenyl]methanamine SMILES: NCC1=C(Cl)C=CC(=C1Cl)C(F)(F)F