accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1092461-19-2

ClBrOFFF

CAS RN 1092461-19-2

IUPAC Name: 2-(bromomethyl)-4-chloro-1-(trifluoromethoxy)benzene
Synonyms: 2-(bromomethyl)-4-chloro-1-(trifluoromethoxy)benzene
Molecular Weight (g/mol): 289.48
Molecular Formula: C8H5BrClF3O
InChi Key: NVZXASWBNNGRKE-UHFFFAOYSA-N
SMILES: FC(F)(F)OC1=C(CBr)C=C(Cl)C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products