man

Test

CAS RN 1093759-51-3

NNI

CAS RN 1093759-51-3

IUPAC Name: 1-benzyl-3-iodo-1H-pyrrolo[2,3-b]pyridine
Synonyms: 1-benzyl-3-iodopyrrolo[2,3-b]pyridine
Molecular Weight (g/mol): 334.16
Molecular Formula: C14H11IN2
InChi Key: KJXCQDCJEMOYCK-UHFFFAOYSA-N
SMILES: IC1=CN(CC2=CC=CC=C2)C2=NC=CC=C12

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results