accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109704-53-2

CH3NCH3CH3H3COBOOHCH3OH3COCH3O

CAS RN 109704-53-2

IUPAC Name: bis(acetyloxy)-λ²-boranuidyl acetate; tetramethylazanium
Synonyms: bis(acetyloxy)-λ²-boranuidyl acetate; tetramethylammonium ion
Molecular Weight (g/mol): 262.09
Molecular Formula: C10H21BNO6
InChi Key: ZWSJBUUORJFGHK-UHFFFAOYSA-N
SMILES: C[N+](C)(C)C.CC(=O)O[B-](OC(C)=O)OC(C)=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products