accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 109826-27-9

H3CNNOHNONOHOOHO(Z)

CAS RN 109826-27-9

IUPAC Name: (2Z)-but-2-enedioic acid; 1-[1-({7-methyl-8-oxa-2-azatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaen-7-yl}methyl)piperidin-4-yl]-2,3-dihydro-1H-1,3-benzodiazol-2-one
Synonyms: 1-[1-({7-methyl-8-oxa-2-azatricyclo[8.4.0.0²,⁶]tetradeca-1(14),3,5,10,12-pentaen-7-yl}methyl)piperidin-4-yl]-3H-1,3-benzodiazol-2-one; maleic acid
Molecular Weight (g/mol): 544.61
Molecular Formula: C30H32N4O6
InChi Key: NGODOSILXOFQPH-BTJKTKAUNA-N
SMILES: OC(=O)\C=C/C(O)=O.CC1(CN2CCC(CC2)N2C(=O)NC3=CC=CC=C23)OCC2=CC=CC=C2N2C=CC=C12
Molecular Weight (g/mol) 
  • (1)
  • (1)
Percent Purity 
  • (1)
  • (1)
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products