accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 110013-18-8

HNHO

CAS RN 110013-18-8

IUPAC Name: (pyrrolidin-3-yl)methanol
Synonyms: pyrrolidin-3-ylmethanol
Molecular Weight (g/mol): 101.15
Molecular Formula: C5H11NO
InChi Key: QOTUIIJRVXKSJU-UHFFFAOYNA-N
SMILES: OCC1CCNC1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products