accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1101854-58-3

NOOOOOHOONOO

CAS RN 1101854-58-3

IUPAC Name: 4-nitrophenyl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Synonyms: 4-nitrophenyl 4-[bis(2H-1,3-benzodioxol-5-yl)(hydroxy)methyl]piperidine-1-carboxylate
Molecular Weight (g/mol): 520.49
Molecular Formula: C27H24N2O9
InChi Key: SEGYOKHGGFKMCX-UHFFFAOYSA-N
SMILES: OC(C1CCN(CC1)C(=O)OC1=CC=C(C=C1)[N+]([O-])=O)(C1=CC2=C(OCO2)C=C1)C1=CC2=C(OCO2)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products