accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1104-22-9

HHClClCH3NNCl

CAS RN 1104-22-9

IUPAC Name: dihydrogen 1-[(4-chlorophenyl)(phenyl)methyl]-4-[(3-methylphenyl)methyl]piperazine dichloride
Synonyms: dihydrogen meclizine dichloride
Molecular Weight (g/mol): 463.87
Molecular Formula: C25H29Cl3N2
InChi Key: VCTHNOIYJIXQLV-UHFFFAOYNA-N
SMILES: [H+].[H+].[Cl-].[Cl-].CC1=CC=CC(CN2CCN(CC2)C(C2=CC=CC=C2)C2=CC=C(Cl)C=C2)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products