accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11042-64-1

H3COOO(S)(R)(S)(R)CH3(R)(S)H3C(S)(R)CH3H3C(R)CH3H3CCH3HO(E)

CAS RN 11042-64-1

IUPAC Name: (1S,3R,6S,8R,11S,12S,15R,16R)-7,7,12,16-tetramethyl-15-[(2R)-6-methylhept-5-en-2-yl]pentacyclo[9.7.0.0¹,³.0³,⁸.0¹²,¹⁶]octadecan-6-yl (2E)-3-(4-hydroxy-3-methoxyphenyl)prop-2-enoate
Synonyms: oryzanol gamma-oryzanol oliver HI-Z 谷维素 gammariza Hi-Z gamma oryzanol gamma-orizanol
Molecular Weight (g/mol): 602.9
Molecular Formula: C40H58O4
InChi Key: FODTZLFLDFKIQH-FSVGXZBPSA-N
SMILES: COC1=CC(\C=C\C(=O)O[C@H]2CC[C@]34C[C@]33CC[C@]5(C)[C@H](CC[C@@]5(C)[C@@H]3CC[C@H]4C2(C)C)[C@H](C)CCC=C(C)C)=CC=C1O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products