accessibility menu, dialog, popup

Test

CAS RN 110488-70-5

CAS RN 110488-70-5

IUPAC Name: (2Z)-3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-(morpholin-4-yl)prop-2-en-1-one
Synonyms: (2Z)-3-(4-chlorophenyl)-3-(3,4-dimethoxyphenyl)-1-(morpholin-4-yl)prop-2-en-1-one
Molecular Weight (g/mol): 387.86
Molecular Formula: C21H22ClNO4
InChi Key: QNBTYORWCCMPQP-JXAWBTAJSA-N
SMILES: COC1=CC=C(C=C1OC)C(=C/C(=O)N1CCOCC1)\C1=CC=C(Cl)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (3)
  • (1)
Concentration 
  • (3)
Solvent or Matrix 
  • (1)
  • (1)
  • (1)
Color 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)