CAS RN 110623-73-9
CAS RN 110623-73-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Epimedin B, MedChemExpress
MedChemExpress Epimedin B, a component extracted from Epimedii Folium, is reported to have antiosteoporotic activity.
