accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111-21-7

H3COOOOOCH3O

CAS RN 111-21-7

IUPAC Name: 2-{2-[2-(acetyloxy)ethoxy]ethoxy}ethyl acetate
Synonyms: ethylenebis(oxyethylene)diacetate
Molecular Weight (g/mol): 234.25
Molecular Formula: C10H18O6
InChi Key: OVOUKWFJRHALDD-UHFFFAOYSA-N
SMILES: CC(=O)OCCOCCOCCOC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products