accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111-67-1

H3CCH3(Z)

CAS RN 111-67-1

IUPAC Name: (2Z)-oct-2-ene
Synonyms: (2Z)-oct-2-ene
Molecular Weight (g/mol): 112.22
Molecular Formula: C8H16
InChi Key: ILPBINAXDRFYPL-HYXAFXHYSA-N
SMILES: CCCCC\C=C/C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products