accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 11104-28-2

A(a)

CAS RN 11104-28-2

IUPAC Name: —
Synonyms: —
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: —
SMILES: —
Solvent or Matrix 
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
Concentration 
  • (4)
  • (2)
Quantity 
  • (2)

Narrow Results

Narrow Results

  • (6)
  • (3)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (4)
  • (2)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products