man

Test

CAS RN 111072-31-2

NNNNOHNOH3COCH3SSOOOOOONNOOOONa

CAS RN 111072-31-2

IUPAC Name: disodium bis(2,3-bis(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)-3H-1,2λ⁵,3,4-tetrazol-2-ylium)
Synonyms: disodium bis(2,3-bis(2-methoxy-4-nitro-5-sulfonatophenyl)-5-(phenylcarbamoyl)-1,2λ⁵,3,4-tetrazol-2-ylium)
Molecular Weight (g/mol): 1347.03
Molecular Formula: C44H32N14Na2O26S4
InChi Key: GUNJHFPTPMSLSP-UHFFFAOYSA-L
SMILES: [Na+].[Na+].COC1=CC(=C(C=C1N1N=C(N=[N+]1C1=CC(=C(C=C1OC)[N+]([O-])=O)S([O-])(=O)=O)C(=O)NC1=CC=CC=C1)S([O-])(=O)=O)[N+]([O-])=O.COC1=CC(=C(C=C1N1N=C(N=[N+]1C1=CC(=C(C=C1OC)[N+]([O-])=O)S([O-])(=O)=O)C(=O)NC1=CC=CC=C1)S([O-])(=O)=O)[N+]([O-])=O

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results