CAS RN 1110781-88-8
CAS RN 1110781-88-8
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
Tocris Bioscienceâ„¢ EPPTB
Trace amine 1 (TA1) receptor antagonist/inverse agonist
2
3