accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1111-67-7

CuSN

CAS RN 1111-67-7

IUPAC Name: λ¹-copper(1+) cyanosulfanide
Synonyms: copper(I) thiocyanate
Molecular Weight (g/mol): 121.62
Molecular Formula: CCuNS
InChi Key: PDZKZMQQDCHTNF-UHFFFAOYSA-M
SMILES: [Cu+].[S-]C#N
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products