accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111150-22-2

CAS RN 111150-22-2

IUPAC Name: tetrasodium 4-{3-[({3,5-bis[(4-sulfonatophenyl)carbamoyl]phenyl}carbamoyl)amino]-5-[(4-sulfonatophenyl)carbamoyl]benzamido}benzene-1-sulfonate
Synonyms: tetrasodium 4-{3-[({3,5-bis[(4-sulfonatophenyl)carbamoyl]phenyl}carbamoyl)amino]-5-[(4-sulfonatophenyl)carbamoyl]benzamido}benzenesulfonate
Molecular Weight (g/mol): 1096.9
Molecular Formula: C41H28N6Na4O17S4
InChi Key: AQJHZNCSXLBXMY-UHFFFAOYSA-J
SMILES: [Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(NC(=O)C2=CC(=CC(NC(=O)NC3=CC(=CC(=C3)C(=O)NC3=CC=C(C=C3)S([O-])(=O)=O)C(=O)NC3=CC=C(C=C3)S([O-])(=O)=O)=C2)C(=O)NC2=CC=C(C=C2)S([O-])(=O)=O)C=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products