CAS RN 1112-54-5
IUPAC Namn:
ethenyltriethylsilane
Synonymer:
ethenyltriethylsilane
Molekylvikt (g/mol):
142.32
Molekylär sammansättning:
C8H18Si
InChi Key:
HBWGDHDXAMFADB-UHFFFAOYSA-N
SMILES:
CC[Si](CC)(CC)C=C
1
–
1
av
1
Resultat
1
Vinyltrietylsilan, 97 %
CAS: 1112-54-5 Molekylformel: C8H18Si Molekylvikt (g/mol): 142.32 MDL-nummer: MFCD00041345 InChI-nyckel: HBWGDHDXAMFADB-UHFFFAOYSA-N Synonym: vinyltriethylsilane,triethylvinylsilane,triethyl vinyl silane,vinyl triethyl silane,ethenyl-triethylsilane,triethylsilyl ethylene,ethenyl triethyl silane,pubchem13130,triethyl vinyl silane # PubChem CID: 559195 IUPAC-namn: etenyl(trietyl)silan LEDER: CC[Si](CC)(CC)C=C