accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1112-56-7

CH2SnH2CH2CH2C

CAS RN 1112-56-7

IUPAC Name: tetraethenylstannane
Synonyms: tetraethenylstannane
Molecular Weight (g/mol): 226.89
Molecular Formula: C8H12Sn
InChi Key: MZIYQMVHASXABC-UHFFFAOYSA-N
SMILES: C=C[Sn](C=C)(C=C)C=C
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products