CAS RN 1112-66-9
CAS RN 1112-66-9
IUPAC Name:
tetrakis(prop-2-en-1-yl)silane
Synonyms:
tetrakis(prop-2-en-1-yl)silane
Molecular Weight (g/mol):
192.38
Molecular Formula:
C12H20Si
InChi Key:
AKRQMTFHUVDMIL-UHFFFAOYSA-N
SMILES:
C=CC[Si](CC=C)(CC=C)CC=C
1
–
1
of
1
results
1
Thermo Scientific Alfa Aesar Tetraallylsilane, 97%
CAS: 1112-66-9 Molecular Formula: C12H20Si Molecular Weight (g/mol): 192.377 MDL Number: MFCD00048161 InChI Key: AKRQMTFHUVDMIL-UHFFFAOYSA-N Synonym: tetraallylsilane,silane, tetra-2-propenyl,tetrakis prop-2-en-1-yl silane,tetrakis prop-2-enyl silane,silane, tetraallyl,tetraallyl silane,zlchem 610,acmc-20ajk5,tetra 2-propenyl silane,silane,tetra-2-propen-1-yl PubChem CID: 565641 IUPAC Name: tetrakis(prop-2-enyl)silane SMILES: C=CC[Si](CC=C)(CC=C)CC=C