accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1117-71-1

CH3OOBr(E)

CAS RN 1117-71-1

IUPAC Name: methyl (2E)-4-bromobut-2-enoate
Synonyms: methyl (2E)-4-bromobut-2-enoate
Molecular Weight (g/mol): 179.01
Molecular Formula: C5H7BrO2
InChi Key: RWIKCBHOVNDESJ-NSCUHMNNSA-N
SMILES: COC(=O)\C=C\CBr
Boiling Point 
  • (3)
  • (3)
Grade 
  • (6)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (3)
  • (3)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products