accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111787-86-1

CH3OH3COHO

CAS RN 111787-86-1

IUPAC Name: 2-methyl-5-(4-methylphenyl)furan-3-carboxylic acid
Synonyms: 2-methyl-5-(4-methylphenyl)furan-3-carboxylic acid
Molecular Weight (g/mol): 216.24
Molecular Formula: C13H12O3
InChi Key: WWJLHTFFOPYRMV-UHFFFAOYSA-N
SMILES: CC1=C(C=C(O1)C1=CC=C(C)C=C1)C(O)=O
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products