accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1118-71-4

H3CCH3H3COOCH3CH3H3C

CAS RN 1118-71-4

IUPAC Name: 2,2,6,6-tetramethylheptane-3,5-dione
Synonyms: 2,2,6,6-tetramethylheptane-3,5-dione
Molecular Weight (g/mol): 184.28
Molecular Formula: C11H20O2
InChi Key: YRAJNWYBUCUFBD-UHFFFAOYSA-N
SMILES: CC(C)(C)C(=O)CC(=O)C(C)(C)C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (2)
  • (2)
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products