accessibility menu, dialog, popup

UserName

The Ultimate Lab Appliances Promotion

Save Up to 78%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1118807-13-8

H3COOHNNSClOCH3

CAS RN 1118807-13-8

IUPAC Name: N-(6-chloro-1,3-benzothiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide
Synonyms: N-(6-chloro-1,3-benzothiazol-2-yl)-3-(3,4-dimethoxyphenyl)propanamide
Molecular Weight (g/mol): 376.86
Molecular Formula: C18H17ClN2O3S
InChi Key: LXFKEVQQSKQXPR-UHFFFAOYSA-N
SMILES: COC1=C(OC)C=C(CCC(=O)NC2=NC3=CC=C(Cl)C=C3S2)C=C1
Grade 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products