accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111969-64-3

CH3OOOHHOH3CCH3

CAS RN 111969-64-3

IUPAC Name: 5-methyl-2-(propan-2-yl)cyclohexyl 2,2-dihydroxyacetate
Synonyms: 2-isopropyl-5-methylcyclohexyl 2,2-dihydroxyacetate
Molecular Weight (g/mol): 230.3
Molecular Formula: C12H22O4
InChi Key: BWZMJRSMHQDFIT-UHFFFAOYNA-N
SMILES: CC(C)C1CCC(C)CC1OC(=O)C(O)O
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
1x1 image pixel for data collection

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products