accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111988-49-9

ClNSNNN(E/Z)

CAS RN 111988-49-9

IUPAC Name: ({3-[(6-chloropyridin-3-yl)methyl]-1,3-thiazolidin-2-ylidene}amino)formonitrile
Synonyms: thiacloprid calypso calypso 70WG
Molecular Weight (g/mol): 252.72
Molecular Formula: C10H9ClN4S
InChi Key: HOKKPVIRMVDYPB-UHFFFAOYSA-N
SMILES: ClC1=CC=C(CN2CCSC2=NC#N)C=N1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products