accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 111991-09-4

CH3OOCH3NHNOHNSOONONCH3H3CN

CAS RN 111991-09-4

IUPAC Name: 2-({[(4,6-dimethoxypyrimidin-2-yl)carbamoyl]amino}sulfonyl)-N,N-dimethylpyridine-3-carboxamide
Synonyms: nicosulfuron
Molecular Weight (g/mol): 410.41
Molecular Formula: C15H18N6O6S
InChi Key: RTCOGUMHFFWOJV-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=NC(NC(=O)NS(=O)(=O)C2=NC=CC=C2C(=O)N(C)C)=N1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products