accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112-42-5

H3COH

CAS RN 112-42-5

IUPAC Name: undecan-1-ol
Synonyms: undecanol
Molecular Weight (g/mol): 172.31
Molecular Formula: C11H24O
InChi Key: KJIOQYGWTQBHNH-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products