accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112-44-7

H3CO

CAS RN 112-44-7

IUPAC Name: undecanal
Synonyms: undecylenyl alcohol
Molecular Weight (g/mol): 170.3
Molecular Formula: C11H22O
InChi Key: KMPQYAYAQWNLME-UHFFFAOYSA-N
SMILES: CCCCCCCCCCC=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products