accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112-45-8

H2CO

CAS RN 112-45-8

IUPAC Name: undec-10-enal
Synonyms: undecylenal
Molecular Weight (g/mol): 168.28
Molecular Formula: C11H20O
InChi Key: OFHHDSQXFXLTKC-UHFFFAOYSA-N
SMILES: C=CCCCCCCCCC=O
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products