accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112-58-3

H3COCH3

CAS RN 112-58-3

IUPAC Name: 1-(hexyloxy)hexane
Synonyms: di-N-hexyl ether
Molecular Weight (g/mol): 186.34
Molecular Formula: C12H26O
InChi Key: BPIUIOXAFBGMNB-UHFFFAOYSA-N
SMILES: CCCCCCOCCCCCC
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products