accessibility menu, dialog, popup

Test

CAS RN 1120-72-5

CAS RN 1120-72-5

IUPAC Name: 2-methylcyclopentan-1-one
Synonyms: cyclopentanone, 2-methyl-
Molecular Weight (g/mol): 98.15
Molecular Formula: C6H10O
InChi Key: ZIXLDMFVRPABBX-UHFFFAOYNA-N
SMILES: CC1CCCC1=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products

Narrow Results