accessibility menu, dialog, popup

UserName

Hard Science, Easy Bioreagents

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1121-66-0

O

CAS RN 1121-66-0

IUPAC Name: cyclohept-2-en-1-one
Synonyms: cyclohept-2-en-1-one
Molecular Weight (g/mol): 110.16
Molecular Formula: C7H10O
InChi Key: WZCRDVTWUYLPTR-UHFFFAOYSA-N
SMILES: O=C1CCCCC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products