accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1122-96-9

H3CONO

CAS RN 1122-96-9

IUPAC Name: 4-methoxypyridin-1-ium-1-olate
Synonyms: 4-methoxypyridin-1-ium-1-olate
Molecular Weight (g/mol): 125.13
Molecular Formula: C6H7NO2
InChi Key: BOFAIBPJCWFJFT-UHFFFAOYSA-N
SMILES: COC1=CC=[N+]([O-])C=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products