accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112246-73-8

(S)B(S)Cl(R)(S)(S)(S)(S)(R)H3CH3CCH3CH3H3CCH3

CAS RN 112246-73-8

IUPAC Name: chlorobis({2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl})borane
Synonyms: chlorobis({2,6,6-trimethylbicyclo[3.1.1]heptan-3-yl})borane
Molecular Weight (g/mol): 320.75
Molecular Formula: C20H34BCl
InChi Key: PSEHHVRCDVOTID-UHFFFAOYNA-N
SMILES: CC1C2CC(CC1B(Cl)C1CC3CC(C1C)C3(C)C)C2(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products