accessibility menu, dialog, popup

UserName

Nurture Your Progress - Savings to support your research

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112410-23-8

CH3OHNNOCH3CH3H3CCH3CH3

CAS RN 112410-23-8

IUPAC Name: N-tert-butyl-N'-(4-ethylbenzoyl)-3,5-dimethylbenzohydrazide
Synonyms: tebufenozide mimic brand of tebufenozide
Molecular Weight (g/mol): 352.48
Molecular Formula: C22H28N2O2
InChi Key: QYPNKSZPJQQLRK-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C=C1)C(=O)NN(C(=O)C1=CC(C)=CC(C)=C1)C(C)(C)C
Molecular Weight (g/mol) 
  • (1)
  • (2)
Concentration 
  • (1)
  • (1)
Solvent or Matrix 
  • (2)
Grade 
  • (1)
Industry Type 
  • (1)

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products