accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1124329-14-1

CH3NOHNNNNONH3COCH3

CAS RN 1124329-14-1

IUPAC Name: 9-cyclopentyl-2-({2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]phenyl}amino)-7-methyl-8,9-dihydro-7H-purin-8-one
Synonyms: 9-cyclopentyl-2-({2-methoxy-4-[(1-methylpiperidin-4-yl)oxy]phenyl}amino)-7-methylpurin-8-one
Molecular Weight (g/mol): 452.56
Molecular Formula: C24H32N6O3
InChi Key: YUKWVHPTFRQHMF-UHFFFAOYSA-N
SMILES: COC1=C(NC2=NC=C3N(C)C(=O)N(C4CCCC4)C3=N2)C=CC(OC2CCN(C)CC2)=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (3)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products