accessibility menu, dialog, popup

UserName

Discover Our Special Offers

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 112539-09-0

CAS RN 112539-09-0

IUPAC Name: 4-(2-oxopyrrolidin-1-yl)benzene-1-sulfonyl chloride
Synonyms: 4-(2-oxopyrrolidin-1-yl)benzenesulfonyl chloride
Molecular Weight (g/mol): 259.7
Molecular Formula: C10H10ClNO3S
InChi Key: OUAVTZYGHGCCCB-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=C(C=C1)N1CCCC1=O
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products