accessibility menu, dialog, popup

UserName

Great Deals on Sample Preparation, Filtration and Storage 

Save Up to 80%

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Applications
Cell Biology
Applications
Chemistry
Genomics
Applications
Production & Bioprocessing
Applications
Production & Bioprocessing
Proteomics
Applications
Main Menu
Documents and Certificates

CAS RN 1125758-85-1

H3CNNHNNHN(E/Z)

CAS RN 1125758-85-1

IUPAC Name: N'-cyano-N''-(1-phenylethyl)-N-(quinolin-5-yl)guanidine
Synonyms: N'-cyano-N''-(1-phenylethyl)-N-(quinolin-5-yl)guanidine
Molecular Weight (g/mol): 315.38
Molecular Formula: C19H17N5
InChi Key: PQYCRDPLPKGSME-UHFFFAOYNA-N
SMILES: CC(N=C(NC#N)NC1=C2C=CC=NC2=CC=C1)C1=CC=CC=C1

Filtered Search Results

Product from some of our suppliers do not display in Filtered Search results. Please clear all filters to see these products